The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024169.3 | NP_077131.2 | 5 - 6 | 501 | 66 |
GenBank mRNA | AK003331.1 | - | 5 - 6 | 502 | 66 |
AK007813.1 | - | 5 - 6 | 465 | 66 | |
AK019132.1 | - | 5 - 6 | 443 | 66 | |
BC002311.1 | - | 6 - 7 | 451 | 66 | |
BC022900.1 | - | 3 - 4 | 1070 | 66 | |
BC037596.1 | - | 5 - 6 | 457 | 66 |