The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024230.2 | NP_077192.1 | 1 - 2 | 155 | 93 |
GenBank mRNA | AK003989.1 | - | 1 - 2 | 58 | 93 |
BC002317.1 | - | 1 - 2 | 155 | 93 |