Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phf10
Entrez Gene ID
72057
Gene Name
PHD finger protein 10
Gene Aliases
1810055P05Rik, AV024533
Chromosome Location
Chr.17: 14944994 - 14961638 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024250.4 NP_077212.3 5 - 6 605 89
GenBank mRNA AB221576.1 - 3 - 4 272 89
AK007873.1 - 5 - 6 585 89
BC002206.1 - 3 - 4 339 89
CA322732.1 - 2 - 3 163 89
CB247892.1 - 5 - 6 605 89

View Full Product Details