The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146307.1 | NP_001139779.1 | 12 - 13 | 1352 | 97 |
NM_024412.3 | NP_077723.3 | 13 - 14 | 1356 | 97 | |
GenBank mRNA | AF124848.1 | - | 12 - 13 | 1449 | 97 |
AY373832.1 | - | 12 - 13 | 1307 | 97 | |
BC037077.1 | - | 13 - 14 | 1360 | 97 |