The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025780.3 | NP_080056.1 | 1 - 2 | 232 | 87 |
GenBank mRNA | AK009162.1 | - | 1 - 2 | 211 | 87 |
AK018577.1 | - | 1 - 2 | 212 | 87 | |
AK081303.1 | - | 1 - 2 | 233 | 87 | |
BC027502.1 | - | 1 - 2 | 140 | 87 |