The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025812.2 | NP_080088.1 | 9 - 10 | 1133 | 87 |
GenBank mRNA | AK004596.1 | - | 9 - 10 | 1153 | 87 |
AK129010.1 | - | 9 - 10 | 1335 | 87 | |
BC013804.1 | - | 9 - 10 | 1133 | 87 | |
DQ182735.1 | - | 9 - 10 | 1133 | 87 |