Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl13
Entrez Gene ID
67455
Gene Name
kelch-like 13
Gene Aliases
1200009K10Rik, AI451128, Bklhd2, D130072F20Rik, mKIAA1309
Chromosome Location
Chr.X: 23219271 - 23365107 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290476.1 NP_001277405.1 2 - 3 406 75
NM_026167.4 NP_080443.1 4 - 5 425 75
GenBank mRNA AK004677.1 - 4 - 5 397 75
AK122491.1 - 2 - 3 406 75
AK191256.1 - 1 - 2 132 75
BC100393.1 - 3 - 4 390 75

View Full Product Details