The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026178.2 | NP_080454.1 | 4 - 5 | 578 | 103 |
GenBank mRNA | AK004819.1 | - | 4 - 5 | 532 | 103 |
AK011418.1 | - | 4 - 5 | 543 | 103 | |
AK033889.1 | - | 4 - 5 | 598 | 103 | |
AK149704.1 | - | 4 - 5 | 528 | 103 | |
AK158577.1 | - | 4 - 5 | 536 | 103 | |
AK171637.1 | - | 4 - 5 | 532 | 103 | |
AK193751.1 | - | 2 - 3 | 223 | 103 | |
AY424300.1 | - | 4 - 5 | 418 | 103 | |
BC021914.1 | - | 4 - 5 | 578 | 103 | |
BC034117.1 | - | 5 - 6 | 930 | 103 |