The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135149.1 | NP_001128621.1 | 8 - 9 | 1613 | 68 |
NM_001135150.1 | NP_001128622.1 | 8 - 9 | 1763 | 68 | |
NM_026228.5 | NP_080504.3 | 8 - 9 | 1722 | 68 | |
GenBank mRNA | AK014895.1 | - | 8 - 9 | 1726 | 68 |
AK016853.1 | - | 8 - 9 | 1500 | 68 | |
AK028215.1 | - | 8 - 9 | 1511 | 68 | |
AK089060.1 | - | 7 - 8 | 1622 | 68 | |
BC006731.1 | - | 7 - 8 | 1424 | 68 |