Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tctn3
Entrez Gene ID
67590
Gene Name
tectonic family member 3
Gene Aliases
4930521E07Rik, AI197391, Tect3
Chromosome Location
Chr.19: 40596446 - 40612281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026260.2 NP_080536.2 1 - 2 292 74
GenBank mRNA AK015861.1 - 1 - 2 280 74
AK016982.1 - 1 - 2 278 74
AK030156.1 - 1 - 2 278 74
BC027335.1 - 1 - 2 292 74
BC080805.1 - 1 - 2 267 74
DQ278871.1 - 1 - 2 309 74

View Full Product Details