Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Necap1
Entrez Gene ID
67602
Gene Name
NECAP endocytosis associated 1
Gene Aliases
1200016B17Rik, AI747569, mFLJ00061
Chromosome Location
Chr.6: 122874557 - 122888941 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026267.2 NP_080543.2 7 - 8 785 78
GenBank mRNA AK004805.1 - 7 - 8 786 78
AK005263.1 - 7 - 8 776 78
AK081598.1 - 7 - 8 786 78
AK168500.1 - 7 - 8 786 78
AK188487.1 - 1 - 2 73 78
AK220236.1 - 7 - 8 771 78
BC011466.1 - 7 - 8 784 78
BC119133.1 - 7 - 8 797 78
BC120841.1 - 7 - 8 868 78

View Full Product Details