Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reg4
Entrez Gene ID
67709
Gene Name
regenerating islet-derived family, member 4
Gene Aliases
2010002L15Rik, GISP, RELP
Chromosome Location
Chr.3: 98222156 - 98236748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026328.2 NP_080604.2 4 - 5 464 79
GenBank mRNA AK008049.1 - 4 - 5 466 79
AK008438.1 - 4 - 5 467 79
AK160329.1 - 4 - 5 462 79
AK162316.1 - 4 - 5 465 79
BC019465.1 - 4 - 5 447 79

View Full Product Details