The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026328.2 | NP_080604.2 | 5 - 6 | 568 | 69 |
GenBank mRNA | AK008049.1 | - | 5 - 6 | 570 | 69 |
AK008438.1 | - | 5 - 6 | 571 | 69 | |
AK160329.1 | - | 5 - 6 | 566 | 69 | |
AK162316.1 | - | 5 - 6 | 569 | 69 | |
BC019465.1 | - | 5 - 6 | 551 | 69 |