The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026334.3 | NP_080610.1 | 7 - 8 | 860 | 87 |
GenBank mRNA | AK009359.1 | - | 7 - 8 | 862 | 87 |
AK009413.1 | - | 7 - 8 | 861 | 87 | |
AK009428.1 | - | 7 - 8 | 864 | 87 | |
AK009437.1 | - | 7 - 8 | 862 | 87 | |
AK009459.1 | - | 7 - 8 | 862 | 87 | |
AK009473.1 | - | 7 - 8 | 861 | 87 | |
AK009474.1 | - | 7 - 8 | 862 | 87 | |
AK009479.1 | - | 7 - 8 | 859 | 87 | |
AK009523.1 | - | 7 - 8 | 862 | 87 | |
AK009525.1 | - | 7 - 8 | 862 | 87 | |
AK009537.1 | - | 7 - 8 | 862 | 87 | |
AK009544.1 | - | 7 - 8 | 857 | 87 | |
AK009546.1 | - | 7 - 8 | 865 | 87 | |
AK009560.1 | - | 7 - 8 | 862 | 87 | |
AK009571.1 | - | 7 - 8 | 861 | 87 | |
AK009573.1 | - | 7 - 8 | 865 | 87 | |
AK009729.1 | - | 7 - 8 | 862 | 87 | |
AK009773.1 | - | 7 - 8 | 862 | 87 | |
AK009875.1 | - | 7 - 8 | 862 | 87 | |
AK009923.1 | - | 7 - 8 | 923 | 87 | |
AK009990.1 | - | 7 - 8 | 862 | 87 | |
AK010019.1 | - | 7 - 8 | 863 | 87 | |
AK010026.1 | - | 7 - 8 | 862 | 87 | |
AK010035.1 | - | 7 - 8 | 862 | 87 | |
AK010036.1 | - | 7 - 8 | 861 | 87 | |
AK010058.1 | - | 7 - 8 | 861 | 87 | |
AK010061.1 | - | 7 - 8 | 862 | 87 | |
AK010091.1 | - | 7 - 8 | 860 | 87 | |
AK010093.1 | - | 7 - 8 | 862 | 87 | |
AK010106.1 | - | 7 - 8 | 862 | 87 | |
AK010110.1 | - | 7 - 8 | 865 | 87 | |
AK010123.1 | - | 7 - 8 | 862 | 87 | |
AK010124.1 | - | 6 - 7 | 856 | 87 | |
AK010125.1 | - | 7 - 8 | 862 | 87 | |
AK010139.1 | - | 7 - 8 | 862 | 87 | |
AK010148.1 | - | 7 - 8 | 862 | 87 | |
AK010173.1 | - | 7 - 8 | 862 | 87 | |
AK010203.1 | - | 7 - 8 | 863 | 87 | |
AK010231.1 | - | 7 - 8 | 861 | 87 | |
AK010236.1 | - | 7 - 8 | 863 | 87 | |
AK075913.1 | - | 7 - 8 | 862 | 87 | |
BC061067.1 | - | 7 - 8 | 860 | 87 |