Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yipf4
Entrez Gene ID
67864
Gene Name
Yip1 domain family, member 4
Gene Aliases
2310034L04Rik
Chromosome Location
Chr.17: 74489493 - 74500277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026417.4 NP_080693.2 4 - 5 712 141
GenBank mRNA AK017721.1 - 4 - 5 690 141
AK017722.1 - 4 - 5 690 141
AK083301.1 - 4 - 5 692 141
AK159227.1 - 4 - 5 673 141
AK166347.1 - 4 - 5 700 141
BC003936.1 - 5 - 6 887 141

View Full Product Details