The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026438.4 | NP_080714.2 | 5 - 6 | 494 | 72 |
GenBank mRNA | AK008575.1 | - | 5 - 6 | 495 | 72 |
AK020291.1 | - | 5 - 6 | 72 | ||
AK088397.1 | - | 5 - 6 | 490 | 72 | |
AK145640.1 | - | 5 - 6 | 544 | 72 | |
AK146300.1 | - | 5 - 6 | 457 | 72 | |
AK151512.1 | - | 5 - 6 | 485 | 72 | |
AK219020.1 | - | 2 - 3 | 153 | 72 | |
BC010468.1 | - | 5 - 6 | 428 | 72 | |
CT010203.1 | - | 5 - 6 | 387 | 72 |