The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026487.3 | NP_080763.2 | 9 - 10 | 1016 | 75 |
GenBank mRNA | AK007147.1 | - | 5 - 6 | 473 | 75 |
AK009419.1 | - | 5 - 6 | 604 | 75 | |
AK014967.1 | - | 9 - 10 | 1033 | 75 | |
AK030719.1 | - | 9 - 10 | 1036 | 75 | |
AK033639.1 | - | 9 - 10 | 1040 | 75 | |
AK150469.1 | - | 9 - 10 | 1036 | 75 | |
AK152059.1 | - | 9 - 10 | 1036 | 75 | |
AK165953.1 | - | 9 - 10 | 1051 | 75 | |
BC029085.1 | - | 9 - 10 | 1016 | 75 | |
BC043051.1 | - | 9 - 10 | 1001 | 75 |