Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hormad1
Entrez Gene ID
67981
Gene Name
HORMA domain containing 1
Gene Aliases
4921522K05Rik, Nohma
Chromosome Location
Chr.3: 95559640 - 95587940 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026489.3 NP_080765.1 1 - 2 51 54
GenBank mRNA AK016743.1 - 1 - 2 54 54

View Full Product Details