Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina12
Entrez Gene ID
68054
Gene Name
serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12
Gene Aliases
4632419J12Rik
Chromosome Location
Chr.12: 104028769 - 104044443 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026535.2 NP_080811.1 3 - 4 1104 70
GenBank mRNA AK014346.1 - 3 - 4 1046 70
AK014589.1 - 3 - 4 1104 70
AY326419.1 - 2 - 3 937 70
BC062143.1 - 3 - 4 1058 70

View Full Product Details