The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026542.3 | NP_080818.1 | 5 - 6 | 463 | 74 |
GenBank mRNA | AK002381.1 | - | 5 - 6 | 421 | 74 |
AK007276.1 | - | 5 - 6 | 442 | 74 | |
AK007934.1 | - | 5 - 6 | 417 | 74 | |
AK010272.1 | - | 5 - 6 | 442 | 74 | |
AK013834.1 | - | 1 - 2 | 107 | 74 | |
AK019396.1 | - | 5 - 6 | 455 | 74 | |
AK078417.1 | - | 5 - 6 | 455 | 74 | |
AK084116.1 | - | 5 - 6 | 454 | 74 | |
AK160922.1 | - | 5 - 6 | 408 | 74 | |
AK161289.1 | - | 5 - 6 | 455 | 74 | |
BC115661.1 | - | 5 - 6 | 363 | 74 | |
BC115662.1 | - | 5 - 6 | 370 | 74 |