The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026612.3 | NP_080888.1 | 1 - 2 | 132 | 92 |
GenBank mRNA | AK007495.1 | - | 1 - 2 | 108 | 92 |
AK010642.1 | - | 1 - 2 | 139 | 92 | |
AK019008.1 | - | 1 - 2 | 137 | 92 | |
AK028334.1 | - | - | 141 | 92 | |
BC013510.1 | - | 1 - 2 | 142 | 92 |