The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026626.2 | NP_080902.1 | 2 - 3 | 335 | 85 |
GenBank mRNA | AK010880.1 | - | 2 - 3 | 188 | 85 |
AK015157.1 | - | 2 - 3 | 117 | 85 | |
AK018894.1 | - | 2 - 3 | 196 | 85 | |
AK019663.1 | - | 2 - 3 | 128 | 85 | |
AK019802.1 | - | 2 - 3 | 137 | 85 | |
AK029880.1 | - | - | 118 | 85 | |
BC025062.1 | - | 2 - 3 | 335 | 85 |