Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkd1
Entrez Gene ID
93960
Gene Name
naked cuticle 1 homolog (Drosophila)
Gene Aliases
2810434J10Rik, 9030215G15Rik, Nkd
Chromosome Location
Chr.8: 88521344 - 88594887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163660.1 NP_001157132.1 6 - 7 646 60
NM_027280.3 NP_081556.3 8 - 9 856 60
GenBank mRNA AF343352.1 - 8 - 9 693 60
AF358134.1 - 8 - 9 772 60
AK166564.1 - 8 - 9 859 60
AK190160.1 - 1 - 2 46 60
BC034838.1 - 8 - 9 830 60
DV662736.1 - 6 - 7 626 60

View Full Product Details