Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec2g
Entrez Gene ID
70809
Gene Name
C-type lectin domain family 2, member g
Gene Aliases
4632413B12Rik, Clr-c, Clr-d, Clrc, Clrd, Clrx, Ddv10, Ocilrp1
Chromosome Location
Chr.6: 128934381 - 128984707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027562.4 NP_081838.1 1 - 2 103 90
GenBank mRNA AK014570.1 - 1 - 2 104 90
BC117543.1 - 1 - 2 93 90

View Full Product Details