Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhrs2
Entrez Gene ID
71412
Gene Name
dehydrogenase/reductase member 2
Gene Aliases
5430405K24Rik, Hep27
Chromosome Location
Chr.14: 55124154 - 55241440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027790.2 NP_082066.2 2 - 3 147 60
GenBank mRNA AK017274.1 - 2 - 3 149 60
BC117731.2 - 1 - 2 40 60

View Full Product Details