The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027910.3 | NP_082186.2 | 1 - 2 | 236 | 101 |
GenBank mRNA | AB053465.1 | - | 1 - 2 | 107 | 101 |
AJ278127.1 | - | 1 - 2 | 103 | 101 | |
AK004964.1 | - | 1 - 2 | 213 | 101 | |
AK079082.1 | - | 1 - 2 | 110 | 101 | |
AK170623.1 | - | 1 - 2 | 108 | 101 | |
BC018154.1 | - | 1 - 2 | 205 | 101 | |
CF169399.1 | - | 1 - 2 | 236 | 101 |