Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc3
Entrez Gene ID
71765
Gene Name
kelch domain containing 3
Gene Aliases
1300011D16Rik, Peas
Chromosome Location
Chr.17: 46674550 - 46680930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163729.2 NP_001157201.2 5 - 6 812 84
NM_027910.3 NP_082186.2 6 - 7 965 84
GenBank mRNA AB053465.1 - 6 - 7 836 84
AJ278127.1 - 6 - 7 832 84
AK004964.1 - 6 - 7 942 84
AK141673.1 - 5 - 6 797 84
AK170623.1 - 6 - 7 837 84
BC018154.1 - 6 - 7 934 84

View Full Product Details