Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkd2
Entrez Gene ID
72293
Gene Name
naked cuticle 2 homolog (Drosophila)
Gene Aliases
2210403L10Rik, AW212591
Chromosome Location
Chr.13: 73818529 - 73847631 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028186.4 NP_082462.3 2 - 3 270 93
GenBank mRNA AF358136.1 - 2 - 3 220 93
AK008814.1 - 2 - 3 252 93
AK145155.1 - 2 - 3 252 93
AK146667.1 - 2 - 3 252 93
AK158350.1 - 2 - 3 252 93
BC019952.1 - 2 - 3 239 93

View Full Product Details