Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkd2
Entrez Gene ID
72293
Gene Name
naked cuticle 2 homolog (Drosophila)
Gene Aliases
2210403L10Rik, AW212591
Chromosome Location
Chr.13: 73818529 - 73847631 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028186.4 NP_082462.3 8 - 9 885 78
GenBank mRNA AF358136.1 - 8 - 9 835 78
AK008814.1 - 8 - 9 867 78
AK081904.1 - 7 - 8 831 78
AK145155.1 - 8 - 9 867 78
AK146667.1 - 8 - 9 867 78
AK158350.1 - 8 - 9 867 78

View Full Product Details