Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35a5
Entrez Gene ID
74102
Gene Name
solute carrier family 35, member A5
Gene Aliases
1010001J06Rik, AU021179, BB097433, D16Ertd450e, D730043G07Rik
Chromosome Location
Chr.16: 45139573 - 45158673 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028756.4 NP_083032.2 4 - 5 706 132
GenBank mRNA AK003124.1 - 4 - 5 634 132
AK030566.1 - 4 - 5 631 132
AK033921.1 - 4 - 5 673 132
AK085567.1 - 4 - 5 645 132
AK150825.1 - 4 - 5 697 132
AK151484.1 - 4 - 5 697 132
AK170026.1 - 4 - 5 673 132
BC011115.1 - 4 - 5 635 132

View Full Product Details