Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Haus8
Entrez Gene ID
76478
Gene Name
4HAUS augmin-like complex, subunit 8
Gene Aliases
2410004L22Rik, Hice1
Chromosome Location
Chr.8: 71251124 - 71272590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029621.3 NP_083897.2 4 - 5 492 79
GenBank mRNA AK010398.1 - 4 - 5 280 79

View Full Product Details