The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030678.3 | NP_109603.2 | 14 - 15 | 2020 | 59 |
GenBank mRNA | AK050813.1 | - | 12 - 13 | 1453 | 59 |
AK169344.1 | - | 14 - 15 | 2016 | 59 | |
AK170404.1 | - | 14 - 15 | 2022 | 59 | |
AK170585.1 | - | 14 - 15 | 1940 | 59 | |
AK171148.1 | - | 14 - 15 | 1975 | 59 | |
AK171426.1 | - | 14 - 15 | 1999 | 59 | |
BC019389.1 | - | 14 - 15 | 1813 | 59 | |
BC131687.1 | - | 14 - 15 | 1867 | 59 | |
BC131688.1 | - | 14 - 15 | 1867 | 59 | |
BC152550.1 | - | 14 - 15 | 1867 | 59 | |
U53218.1 | - | 14 - 15 | 1858 | 59 | |
X94616.1 | - | 14 - 15 | 1867 | 59 |