The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030692.4 | NP_109617.1 | 5 - 6 | 497 | 73 |
GenBank mRNA | AJ245720.1 | - | 5 - 6 | 627 | 73 |
AJ297526.1 | - | 5 - 6 | 498 | 73 | |
AK089174.1 | - | 5 - 6 | 581 | 73 | |
AK122390.1 | - | 5 - 6 | 562 | 73 | |
AK154759.1 | - | 5 - 6 | 597 | 73 | |
AK161111.1 | - | 5 - 6 | 531 | 73 | |
AK171080.1 | - | 5 - 6 | 581 | 73 | |
BC117752.1 | - | 5 - 6 | 498 | 73 | |
BC117753.1 | - | 5 - 6 | 498 | 73 |