The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030703.2 | NP_109628.1 | 2 - 3 | 653 | 66 |
GenBank mRNA | AB021969.1 | - | 2 - 3 | 630 | 66 |
AF326477.1 | - | 2 - 3 | 653 | 66 | |
BC014692.1 | - | 2 - 3 | 621 | 66 |