Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn1r63
Entrez Gene ID
81017
Gene Name
vomeronasal 1 receptor 63
Gene Aliases
V1R1, V1rd1, V3R1
Chromosome Location
Chr.7: 5802345 - 5805445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030742.1 NP_109667.1 1 - 2 143 79
GenBank mRNA AF324866.1 - 1 - 2 143 79

View Full Product Details