Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klb
Entrez Gene ID
83379
Gene Name
klotho beta
Gene Aliases
AV071179, betaKlotho
Chromosome Location
Chr.5: 65348411 - 65384020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031180.2 NP_112457.1 2 - 3 1333 87
GenBank mRNA AF165170.1 - 2 - 3 1333 87
AF178429.1 - 2 - 3 1328 87
AK133823.1 - 2 - 3 1360 87
BC138008.1 - 2 - 3 1422 87
BC138010.1 - 2 - 3 1422 87

View Full Product Details