Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap4
Entrez Gene ID
83383
Gene Name
transcription factor AP4
Gene Aliases
AI642933, AP-4, D930048N17Rik, Tcfap4, bHLHc41
Chromosome Location
Chr.16: 4544661 - 4560772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031182.2 NP_112459.1 6 - 7 1026 66
GenBank mRNA AF161262.1 - 6 - 7 1026 66
AK053091.1 - 6 - 7 818 66
AK166424.1 - 6 - 7 1165 66
BC047270.1 - 6 - 7 933 66
BC054777.1 - 6 - 7 917 66

View Full Product Details