Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kel
Entrez Gene ID
23925
Gene Name
Kell blood group
Gene Aliases
CD238, Ece3
Chromosome Location
Chr.6: 41686329 - 41704334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032540.3 NP_115929.3 18 - 19 2236 86
GenBank mRNA AF252870.1 - 18 - 19 2208 86
AI894156.1 - 1 - 2 86
AK028430.1 - 18 - 19 2210 86
BC099961.1 - 18 - 19 2234 86

View Full Product Details