Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
M1ap
Entrez Gene ID
110958
Gene Name
meiosis 1 associated protein
Gene Aliases
D6Mm5e
Chromosome Location
Chr.6: 82946908 - 83030309 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033079.2 NP_149070.2 2 - 3 330 70
GenBank mRNA AF084364.1 - 1 - 2 299 70
AK131954.1 - 2 - 3 330 70
BC119803.1 - 1 - 2 257 70
BC122521.1 - 1 - 2 262 70

View Full Product Details