Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubqln4
Entrez Gene ID
94232
Gene Name
ubiquilin 4
Gene Aliases
A1Up, A1u, AI663987, CIP75, UBIN
Chromosome Location
Chr.3: 88553677 - 88569725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033526.2 NP_277068.1 7 - 8 1341 55
GenBank mRNA AB040050.1 - 7 - 8 1341 55
AK150211.1 - 7 - 8 1278 55
AK156515.1 - 7 - 8 1295 55
BC017686.1 - 7 - 8 1303 55

View Full Product Details