Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid1a
Entrez Gene ID
93760
Gene Name
AT rich interactive domain 1A (SWI-like)
Gene Aliases
1110030E03Rik, BAF250a, Osa1, Smarcf1
Chromosome Location
Chr.4: 133679008 - 133753981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080819.1 NP_001074288.1 19 - 20 5129 107
GenBank mRNA AF268912.1 - 18 - 19 3992 107
BC082554.1 - - 4373 107

View Full Product Details