The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033602.2 | NP_291080.2 | 5 - 6 | 981 | 113 |
GenBank mRNA | AF302504.1 | - | 5 - 6 | 910 | 113 |
AK030564.1 | - | 5 - 6 | 1028 | 113 | |
AK033815.1 | - | 2 - 3 | 287 | 113 | |
AK045515.1 | - | 4 - 5 | 854 | 113 | |
BC027062.1 | - | 4 - 5 | 820 | 113 |