The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052994.2 | NP_443720.1 | 7 - 8 | 919 | 72 |
GenBank mRNA | AJ278999.1 | - | 7 - 8 | 921 | 72 |
AK122233.1 | - | 7 - 8 | 581 | 72 | |
AK140483.1 | - | 7 - 8 | 826 | 72 | |
AK162941.1 | - | 7 - 8 | 824 | 72 | |
BC057324.1 | - | 7 - 8 | 919 | 72 | |
BC060210.1 | - | 7 - 8 | 871 | 72 |