The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053080.3 | NP_444310.3 | 2 - 3 | 273 | 86 |
GenBank mRNA | AF152359.1 | - | 2 - 3 | 228 | 86 |
AF246711.1 | - | 2 - 3 | 221 | 86 | |
AF253409.1 | - | 2 - 3 | 260 | 86 | |
AF280404.1 | - | 2 - 3 | 225 | 86 | |
AK043530.1 | - | 2 - 3 | 270 | 86 | |
AK086764.1 | - | 2 - 3 | 286 | 86 | |
AK147006.1 | - | 2 - 3 | 274 | 86 | |
BC058277.1 | - | 2 - 3 | 217 | 86 |