The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053146.2 | NP_444376.1 | 1 - 1 | 521 | 126 |
GenBank mRNA | AF326314.1 | - | 1 - 1 | 353 | 126 |
AK134846.1 | - | - | 521 | 126 | |
AY013783.1 | - | 1 - 1 | 353 | 126 | |
BC138121.1 | - | 1 - 1 | 440 | 126 | |
BC138122.1 | - | 1 - 1 | 440 | 126 |