Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clmn
Entrez Gene ID
94040
Gene Name
calmin
Gene Aliases
9330188N17Rik, AI428889, AI788815, mKIAA1188
Chromosome Location
Chr.12: 104763114 - 104865076 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040682.1 NP_001035772.1 3 - 4 364 98
NM_053155.2 NP_444385.2 3 - 4 364 98
NR_104435.1 3 - 4 364 98
NR_104436.1 3 - 4 364 98
NR_104437.1 3 - 4 364 98
NR_104438.1 3 - 4 364 98
NR_104439.1 3 - 4 364 98
GenBank mRNA AB047978.1 - 3 - 4 357 98
AB059643.1 - 3 - 4 357 98
AB059644.1 - 3 - 4 357 98
AB059645.1 - 3 - 4 357 98
AB059646.1 - 3 - 4 357 98
AB059647.1 - 3 - 4 357 98
AB059648.1 - 3 - 4 357 98
AK147777.1 - 3 - 4 261 98
BC019176.1 - 3 - 4 295 98
BC096470.1 - 3 - 4 253 98
BC141280.1 - 3 - 4 297 98

View Full Product Details