The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053201.3 | NP_444431.3 | 1 - 1 | 1948 | 115 |
GenBank mRNA | AF319978.1 | - | 1 - 1 | 1151 | 115 |
AK030032.1 | - | 1 - 1 | 1942 | 115 | |
AK051625.1 | - | 1 - 1 | 1922 | 115 | |
AK081378.1 | - | 1 - 1 | 1924 | 115 | |
BC059039.1 | - | 1 - 1 | 1948 | 115 |