The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197321.1 | NP_001184250.1 | 11 - 12 | 1395 | 69 |
NM_001197322.1 | NP_001184251.1 | 8 - 9 | 1002 | 69 | |
NM_053202.2 | NP_444432.1 | 12 - 13 | 1547 | 69 | |
GenBank mRNA | AB221601.1 | - | 9 - 10 | 1242 | 69 |
AB221602.1 | - | 9 - 10 | 1240 | 69 | |
AB594192.1 | - | 13 - 14 | 1955 | 69 | |
AF339103.1 | - | 9 - 10 | 1236 | 69 | |
AF339104.1 | - | 9 - 10 | 1236 | 69 | |
AF339105.1 | - | - | 1016 | 69 | |
AK033368.1 | - | 6 - 7 | 712 | 69 | |
AK077062.1 | - | 8 - 9 | 897 | 69 | |
BC064764.1 | - | 11 - 12 | 1381 | 69 |