Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxp2
Entrez Gene ID
114142
Gene Name
forkhead box P2
Gene Aliases
2810043D05Rik, AI449000, CAG-16, D0Kist7
Chromosome Location
Chr.6: 14901226 - 15441977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286607.1 NP_001273536.1 11 - 12 1723 90
NM_053242.4 NP_444472.2 13 - 14 2123 90
NM_212435.1 NP_997600.1 11 - 12 1786 90
GenBank mRNA AF339106.1 - 10 - 11 1464 90
AK051668.1 - 13 - 14 2106 90
AK082361.1 - 13 - 14 1795 90
AY079003.1 - 13 - 14 1752 90
BC058960.1 - 11 - 12 1720 90
BC062926.1 - 11 - 12 1786 90

View Full Product Details