Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok4
Entrez Gene ID
114255
Gene Name
docking protein 4
Gene Aliases
-
Chromosome Location
Chr.8: 94863828 - 94876312 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053246.3 NP_444476.1 2 - 3 389 87
GenBank mRNA AF418207.1 - 1 - 2 273 87
AK032352.1 - 2 - 3 379 87
AK043670.1 - 1 - 2 301 87
AK077612.1 - 2 - 3 380 87
AK142621.1 - 1 - 2 296 87
AK156485.1 - 2 - 3 389 87
BC004705.1 - 2 - 3 350 87

View Full Product Details